Acetophenone 99%
New Product
Leverancier: Apollo Scientific
Ga naar:
Over dit artikel
Usage: Organic building block/reagent
Synoniemen: Methyl phenyl ketone, Phenyl methyl ketone
- CAS-nummer:98-86-2
- Formule:C₈H₈O
- MDL-nummer:MFCD00008724
- Molecular Weight:120,15 g/mol
4 Opties Hieronder Beschikbaar
Product Details & Documents
Usage: Organic building block/reagent
Synoniemen: Methyl phenyl ketone, Phenyl methyl ketone
- CAS-nummer:98-86-2
- Formule:C₈H₈O
- MDL-nummer:MFCD00008724
- Molecular Weight:120,15 g/mol
Product Family Options
Productinformatie
- VerpakkingBeschikbaarheidPrice
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product