Selenium(IV) oxide 99,8%
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Inorganic salt or catalyst
- CAS Number:7446-08-4
- Formula:H₂O₂Se
- MDL Number:MFCD00003562
- Molecular Weight:112,97467 g/mol
- UN:3283
2 Options Available Below
Product Details & Documents
Usage: Inorganic salt or catalyst
- CAS Number:7446-08-4
- Formula:H₂O₂Se
- MDL Number:MFCD00003562
- Molecular Weight:112,97467 g/mol
- UN:3283
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



