Orders LinkContactUsLinkComponent
Acetaminophen ≥98%
Acetaminophen ≥98%
Bottle
Catalogus # APOSBUP18863-500MG
New Product
Leverancier:   Apollo Scientific
Acetaminophen ≥98%
Catalogus # APOSBUP18863-500MG
Leverancier:  Apollo Scientific
New Product
CAS-nummer:  103-90-2
Acetaminophen ≥98%
Bottle
CAS-nummer:  103-90-2
Price
52,10 €
Log in voor beschikbaarheid
1 * 500 mg
Product Details & Documents
Usage: Biochemical/life science applications
Synoniemen:  Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
  • CAS-nummer:103-90-2
  • Formule:C₈H₉NO₂
  • Molecular Weight:151,16 g/mol
Specificaties
Specificaties
  • Verpakking:
    Bottle