Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

2-Hydroxycinnamic acid ≥98%

Bottle
Catalogus # APOSBUP18101-500MG
New Product
Leverancier:   Apollo Scientific
2-Hydroxycinnamic acid ≥98%
Catalogus # APOSBUP18101-500MG
Leverancier:  Apollo Scientific
New Product
CAS-nummer:  614-60-8
2-Hydroxycinnamic acid ≥98%

Bottle

CAS-nummer:  614-60-8
Price
53,60 €
Log in voor beschikbaarheid
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synoniemen:  (E)-3-(2-Hydroxyphenyl)acrylic acid, 2-Hydroxycinnamic acid, predominantly trans, trans-2-Hydroxycinnamic acid, trans-o-Coumaric acid, o-Coumaric acid
  • CAS-nummer:614-60-8
  • Formule:C₉H₈O₃
  • Molecular Weight:164,16 g/mol
Specificaties

Specificaties

  • Verpakking:
    Bottle

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.