Hygromycin B ≥98%
New Product
Leverancier: Apollo Scientific
Ga naar:
Over dit artikel
Usage: Biochemical/life science applications
Synoniemen: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
- CAS-nummer:31282-04-9
- Formule:C₂₀H₃₇N₃O₁₃
- Molecular Weight:527,52 g/mol
3 Opties Hieronder Beschikbaar
Product Details & Documents
Usage: Biochemical/life science applications
Synoniemen: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
- CAS-nummer:31282-04-9
- Formule:C₂₀H₃₇N₃O₁₃
- Molecular Weight:527,52 g/mol
Product Family Options
Productinformatie
- VerpakkingBeschikbaarheidPrice
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product



