(2-Hydroxypropyl)-beta-cyclodextrin 98%
New Product
Leverancier: Apollo Scientific
Ga naar:
Over dit artikel
Usage: Biochemical/life science applications
Synoniemen: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
- CAS-nummer:128446-35-5
- Formule:C₆₃H₁₁₂O₄₂
- MDL-nummer:MFCD00069372
- Molecular Weight:1541,54 g/mol
- EINECS:420-920-1
6 Opties Hieronder Beschikbaar
Product Details & Documents
Usage: Biochemical/life science applications
Synoniemen: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
- CAS-nummer:128446-35-5
- Formule:C₆₃H₁₁₂O₄₂
- MDL-nummer:MFCD00069372
- Molecular Weight:1541,54 g/mol
- EINECS:420-920-1
Product Family Options
Productinformatie
- VerpakkingBeschikbaarheidPrice
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product
- Specificaties:New ProductNew Product