Allantoin 98%
New Product
Leverancier: Apollo Scientific
Ga naar:
Over dit artikel
Usage: Organic building block/reagent
Synoniemen: 1-(2,5-Dioxoimidazolidin-4-yl)urea
- CAS-nummer:97-59-6
- Formule:C₄H₆N₄O₃
- Molecular Weight:158,12 g/mol
3 Opties Hieronder Beschikbaar
Product Details & Documents
Usage: Organic building block/reagent
Synoniemen: 1-(2,5-Dioxoimidazolidin-4-yl)urea
- CAS-nummer:97-59-6
- Formule:C₄H₆N₄O₃
- Molecular Weight:158,12 g/mol



