Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Sucrose ≥98%

Sucrose ≥98%

Bottle
Catalogus # APOSBUP18809-500MG
New Product
Leverancier:   Apollo Scientific
Sucrose ≥98%
Catalogus # APOSBUP18809-500MG
Leverancier:  Apollo Scientific
New Product
CAS-nummer:  57-50-1
Sucrose ≥98%

Bottle

CAS-nummer:  57-50-1
Price
37,70 €
Log in voor beschikbaarheid
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synoniemen:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS-nummer:57-50-1
  • Formule:C₁₂H₂₂O₁₁
  • Molecular Weight:342,3 g/mol
Specificaties

Specificaties

  • Verpakking:
    Bottle

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.