Orders LinkContactUsLinkComponent
DIVTECH Fluogreen Small Molecule Delivery Nanoparticles
DIVTECH Fluogreen Small Molecule Delivery Nanoparticles
Fluogreen small molecule delivery nanoparticles (Hydrophobic and amphiphilic small molecules [drug-like APIs]; Adherent and suspension cells, organoids, 3D cultures, bacteria, rodents and zebrafish), 1 ml
Catalogus # DIVSDIV010F1
New Product
Leverancier:   DIVERSA TECHNOLOGIES
DIVTECH Fluogreen Small Molecule Delivery Nanoparticles
Catalogus # DIVSDIV010F1
Leverancier:  DIVERSA TECHNOLOGIES
New Product
CAS-nummer:  
DIVTECH Fluogreen Small Molecule Delivery Nanoparticles
DIVTECH Fluogreen Small Molecule Delivery Nanoparticles
Fluogreen small molecule delivery nanoparticles (Hydrophobic and amphiphilic small molecules [drug-like APIs]; Adherent and suspension cells, organoids, 3D cultures, bacteria, rodents and zebrafish), 1 ml
CAS-nummer:  
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
DIVERSA’s Small molecule delivery nanoparticles simplify the challenge of formulating poorly water soluble compounds. With their high loading efficiency and proven stability in biological environments, they enable researchers to explore drug activity in both simple cell models and complex in vivo systems. Designed for fast, reproducible results, the kit provides flexibility for discovery projects and translational research. Fluorescent nanoparticles streamline visualisation and biodistribution studies.
  • Easy-to-use and ready-to-load nanoparticle formulation
    High loading capacity for hydrophobic and amphiphilic compounds
    Suitable for in vitro and in vivo applications
    Compatible with 2D and 3D cell culture models, including immortalise cells, primary cells and organoids
    Non-toxic and biodegradable components
    Stable in biological media
    No specialised equipment required for preparation
    Ideal for internalisation and biodistribution studies
    Trackable by standard fluorescence detection platforms
Poor water solubility remains a major obstacle in the development of many promising drug candidates, particularly hydrophobic small molecules. While such compounds can dissolve in solvents like DMSO, their limited solubility in biological media restricts their applicability in preclinical models. DIVERSA’s Small molecule delivery nanoparticles address this challenge by encapsulating small molecules into biocompatible, stable carriers that remain functional in serum-containing environments.
The platform provides high loading efficiency for hydrophobic and amphiphilic drugs, enabling researchers to assess pharmacological effects across 2D, 3D, and animal models. The kit is designed for straightforward use without specialized equipment, accelerating experimental workflows. Optional fluorescent versions allow monitoring of nanoparticle distribution, uptake, and release using standard detection methods such as microscopy, flow cytometry, or plate readers. By combining versatility with reproducibility, DIV010F1 empowers researchers to unlock the therapeutic potential of otherwise poorly soluble molecules.
Levering: The kit contains one vial with the lipid mix for formulation. Each kit is provided with 1X DIVTECH empty vial for formulation preparation. Includes two pipette tips for 1 ml pipette to support accurate handling. Lot number and storage conditions indicated on every vial and box.
Let op: Products are intended for research use only (RUO).
Specificaties
Specificaties
  • Omschrijving:
    Fluogreen small molecule delivery nanoparticles (Hydrophobic and amphiphilic small molecules [drug-like APIs]; Adherent and suspension cells, organoids, 3D cultures, bacteria, rodents and zebrafish)
  • Volume:
    1 ml