Orders LinkContactUsLinkComponent
Soder-Wick® Desoldering Braids
ESD-veilige spoelen #5 bruin, het meest geschikt voor aansluitingen; 10 spoelen per hersluitbare zak, 6 zakken; breedte: 3,7 mm; lengte: 1,5 m
Catalogus # TEXWSW18055
New Product
Leverancier:   Texwipe
Soder-Wick® Desoldering Braids
Catalogus # TEXWSW18055
Leverancier:  Texwipe
New Product
Soder-Wick® Desoldering Braids
ESD-veilige spoelen #5 bruin, het meest geschikt voor aansluitingen; 10 spoelen per hersluitbare zak, 6 zakken; breedte: 3,7 mm; lengte: 1,5 m
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Soder-Wick Rosin has the fastest wicking action, but rosin flux leaves behind residues that should be removed with a flux remover. Flux-Off Rosin remover is an ideal choice for your cleaning process.
  • Non corrosive ultra high purity Type R rosin flux
    Minimises the risk of heat damage to the board
    Will not leave ionic contamination on the boards
Soder-Wick brand desoldering wick offers the state of the art in desoldering technology. Soder-Wick is designed for today’s heat sensitive electronic components using lighter mass, pure copper braid construction that allows for better thermal conductivity, even at low temperatures. Soder-Wick responds faster than conventional desoldering braids thereby minimising overheating and preventing PCB damage.
Specifications:
MIL-F-14256 F type R flux
NASA-STD-8739.3 Soldered Electrical Connections
DOD-STD-883E, Method 2022
ANSI/IPC J STD-004, Type ROL0
Packaging: Each 25' or longer spool is individually packaged in a sealed bag. 5' and 10' bobbins have 2 packaging options.
Specificaties
  • Omschrijving:
    ESD-veilige spoelen #5 bruin, het meest geschikt voor aansluitingen; 10 spoelen per hersluitbare zak, 6 zakken; breedte: 3,7 mm; lengte: 1,5 m