Mobile Unit for Modules, E 941
Mobiele unit voor orthopedische instrumenten, roestvrij staal, PG 8527, PG 8528, PLW 8615, PLW 8616, PLW 8617
Catalogus # MIEL12895230
New Product
Irregular Voltage
Leverancier: MIELE
Mobile Unit for Modules, E 941
Catalogus # MIEL12895230
Leverancier: MIELE
New Product
Irregular Voltage
Ga naar:
Product Details & Documents
Mobile unit (E 941) for modules on 2 levels.
Suitable for up to 2 modules per level depending on the size of the load
Coupling of the mobile unit to the water and drying air supply
Large-capacity washer-disinfectors
Reprocessing of laboratory glassware
Magnetic strip for automatic mobile unit recognition (incl. magnets): 1
Specificaties
- Omschrijving:Mobiele unit voor orthopedische instrumenten, roestvrij staal
- Voor:PG 8527, PG 8528, PLW 8615, PLW 8616, PLW 8617
- Cat. No.:MIEL12895230