Absorbent Sheets
Over dit artikel
The Transposafe absorption material is available in various models, sizes and absorption volumes. The absorption sheets offer various application options.
High absorption, suitable for Post NL-approved shipments
Suitable for safely transporting blood and other diagnostic materials
Can be used in the food industry
They can be used for the transport of fluids such as blood and other diagnostic materials that fall under UN3373 regulations. They are also suitable for the food industry, for packaging and transporting meat or fish, among other things.
4 Opties Hieronder Beschikbaar
Product Details & Documents
The Transposafe absorption material is available in various models, sizes and absorption volumes. The absorption sheets offer various application options.
High absorption, suitable for Post NL-approved shipments
Suitable for safely transporting blood and other diagnostic materials
Can be used in the food industry
They can be used for the transport of fluids such as blood and other diagnostic materials that fall under UN3373 regulations. They are also suitable for the food industry, for packaging and transporting meat or fish, among other things.
Product Family Options
Productinformatie
- TypeAbsorptievermogenCapaciteitKleurBeschikbaarheidPrice
- 216-1613Absorberend vel, 210×300 mmAbsorberend vel, 210×300 mm1,5 L (absorptiecapaciteit)witLog in voor beschikbaarheid1 * 375 ST195,00 €Specificaties:New ProductNew Product
- TRSPDF110140Absorberend vel, 110×140 mmAbsorberend vel, 110×140 mm200 ml (absorptiecapaciteit)witLog in voor beschikbaarheid1 * 1.000 ST231,00 €
- TRSPDF225250Absorberend vel, 225×250 mmAbsorberend vel, 225×250 mm1 L (absorptiecapaciteit)witLog in voor beschikbaarheid1 * 1.000 ST543,00 €
- TRSPDF150240BAbsorberend vel, 3 buizen, 185×125 mm + 40 mm flapAbsorberend vel, 3 buizen, 185×125 mm + 40 mm flap225 ml (absorptiecapaciteit)witLog in voor beschikbaarheid1 * 50 ST
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.