Handheld Digital Microscope Edge Plus AM8917MT4A
The AM8917MT4A is a feature-packed microscope integrated with coaxial light and EDOF, answering needs for inspecting reflective surfaces and capturing all-in-focus images under 400x magnification. AM8917MT4A includes advanced features such as DPQ, EDR, AMR, and both auto and manual refocus.
High Quality Optics: The optics adopted in the Edge series reveals the finest details, answering the needs of the most demanding microscopy applications.
8.0 Megapixel: The 8.0-megapixel sensor captures a large field of view with a 16:9 aspect ratio and delivers natural-looking colors and enhanced image contrast at a resolution of up to 3840×2160.
Coaxial-Brightfield Illumination: The coaxial illumination uses a through-the-lens design, offering a compact yet high-quality solution for revealing imperfections and fine details on reflective surfaces.
Enhanced Flexible LED Control (eFLC, with coaxial light): Enhancing FLC capabilities, the eFLC can adjust the light intensity on two assigned groups of LED quadrants and the coaxial light, with 32 and 6 levels of adjustment respectively.
Extended Depth of Field (EDOF): Viewing rough surfaces with height ranges out of depth of focus, the EDOF can take several images at different focuses and stack them automatically within a click.
Extended Dynamic Range (EDR): Observing high contrast or reflective surfaces, the EDR can help to reveal the details of dark or bright areas by stacking images taken at different exposure levels.
Auto and Manual Refocus: Using mouse inputs, the refocusing capability helps fine-tune the focus, either automatically by clicking at the point of interest or manually by scrolling the mouse.
Automatic Magnification Readout (AMR): The Automatic Magnification Readout (AMR) detects the magnification in use, eliminating manual input errors and enhancing measurement accuracy in tasks with frequent magnification adjustments.
Robust Metal Housing: The anodized aluminum alloy housing offers durability and EMI shielding, effectively guarding against impacts, abrasions, and electromagnetic noise.
- Vergroting:415 ~ 460x
- Model:AM8917MT4A
- Resolutie:8.0 Megapixel (3840 x 2160) on DinoCapture (Windows); 6.0 Megapixel (2880 x 2160) on DinoXcope (macOS)
- Omschrijving:Handmicroscoop, 33H×32D mm
- Cat. No.:IDCPAM8917MT4A
- Computervereisten:CPU: 2.9 GHz dual-core Intel Core i5 equivalent or greater; RAM: 4 GB or more; HD: 150 MB available space or more
- Frame rate:10 fps at 8.0 MP / 6.0 MP, MJPEG; 25 fps at 2.0 MP, MJPEG