Orders LinkContactUsLinkComponent
Dispensing Lines
Dispensing Lines
New Product
Supplier:   RAYPA
Dispensing Lines
Supplier:  RAYPA
New Product
Certificates
Dispensing Lines
Dispensing Lines
3 Options Available Below
Product Details & Documents
Additional silicone dosing tubes, with metal nozzle for dispensing sterile culture media.
  • Silicone dosing tubes with press-fit connection and metal nozzle
  • Sterile medium is dispensed through the flame-sterilizable nozzle
  • Made of MVQ silicone; heat-resistant up to 200 °C; 60 A hardness
  • Available in multiple diameters
  • Steam and pressurised air enable easy line cleaning before, during, and after use
  • All models are autoclavable, but autoclaving after each use is unnecessary thanks to the built-in steam cleaning program
Ordering information: Dispensing lines can be customised, adjusting their length or adding modifications to adapt them to the specific production requirements of the customer.
Product Family Options
Product Information
  • Omschrijving
    Availability
    Price
  • Extra sets doseerleidingen TUB-DOSIF-8, met een extra set om te voorkomen dat de doseerleiding stopt in geval van contaminatie, reiniging of breuk; 2 m lang; 8 mm inw. Ø, 11,2 mm uitw. Ø
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,00 €
    51,00 €
    1 * 1 ST
       51,00 €
    Specifications:
    New Product
    New Product
  • Extra sets doseerleidingen, TUB-DOSIF-3,2, hebben een extra set om te voorkomen dat de doseerleiding stopt in geval van verontreiniging, reiniging of breuk; lengte 2 m; 3,2 mm inw.Ø, 6,4 mm uitw.Ø
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,00 €
    22,00 €
    1 * 1 ST
       22,00 €
    Specifications:
    New Product
    New Product
  • Extra sets doseerleidingen, TUB-DOSIF-4, hebben een extra set om te voorkomen dat de doseerleiding stopt in geval van verontreiniging, reiniging of breuk; lengte 2 m; 4 mm inw.Ø, 7,2 mm uitw.Ø
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,00 €
    29,00 €
    1 * 1 ST
       29,00 €
    Specifications:
    New Product