Sharpsafe® Needle Containers
Over dit artikel
Needle containers are made up of more than 10% recycled plastic.
Star disconnector for pen injectors
Combined Luer lock & Luer slip disconnector
Safe assembly - 4 audible clicks
Clearly visible filling limit and safety space
Carrying handle - compatible with all SafeClip holders
Dual Protect ProTM: Optimal filling guide and overfill prevention.
4 Opties Hieronder Beschikbaar
Product Details & Documents
Needle containers are made up of more than 10% recycled plastic.
Star disconnector for pen injectors
Combined Luer lock & Luer slip disconnector
Safe assembly - 4 audible clicks
Clearly visible filling limit and safety space
Carrying handle - compatible with all SafeClip holders
Dual Protect ProTM: Optimal filling guide and overfill prevention.
Product Family Options
Productinformatie
- CapaciteitBeschikbaarheidPrice
- HUARSHA-51025610Sharpsafe® naaldcontainer met dubbele bescherming, NL/FR-labelSharpsafe® naaldcontainer met dubbele bescherming, NL/FR-labelLog in voor beschikbaarheid1 * 48 ST293,00 €Specificaties:New ProductNew Product
- HUARSHA-51025720Sharpsafe® NPD grijs internationaal geel deksel, EN/DE/IT/ES/FR labelSharpsafe® NPD grijs internationaal geel deksel, EN/DE/IT/ES/FR label5 LLog in voor beschikbaarheid1 * 48 ST293,00 €Specificaties:New ProductNew Product
- HUARSHA-51125720Sharpsafe® NPD grijs internationaal geel deksel, EN/DE/IT/ES/FR labelSharpsafe® NPD grijs internationaal geel deksel, EN/DE/IT/ES/FR label7 LLog in voor beschikbaarheid1 * 48 ST387,00 €Specificaties:New ProductNew Product
- HUARSHA-51125610Sharpsafe® naaldcontainer met dubbele bescherming, NL/FR-labelSharpsafe® naaldcontainer met dubbele bescherming, NL/FR-label7 LLog in voor beschikbaarheid1 * 48 ST387,00 €Specificaties:New ProductNew Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.