Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Dithiooxamide Metal chelating agent

Dithiooxamide Metal chelating agent

Catalogus # ABCAAB146204-5
Leverancier:   Abcam
Dithiooxamide Metal chelating agent
Catalogus # ABCAAB146204-5
Leverancier:  Abcam
CAS-nummer:  79-40-3
Dithiooxamide Metal chelating agent
CAS-nummer:  79-40-3
Price
97,40 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents

Chelating agent used in the determination of copper(II), nickel(II) and cobalt(II). sulphur analog of oxamide.

Let op: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synoniemen:  Ethanebis(thioamide), Rubeanic acid
  • CAS-nummer:79-40-3
  • Molecular Weight:120,2 g/mol
Specificaties

Specificatie testresultaten

  • Constituents::
    100% Dithiooxamide
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.