Orders LinkContactUsLinkComponent
10X Blocking Buffer, Abcam
10X Blocking Buffer, Abcam
10X Blocking Buffer
Catalogus # ABCAAB126587-50
Marketsource
Leverancier:   Abcam
10X Blocking Buffer, Abcam
Catalogus # ABCAAB126587-50
Leverancier:  Abcam
Marketsource
CAS-nummer:  
10X Blocking Buffer, Abcam
10X Blocking Buffer
CAS-nummer:  
Price
82,00 €
Log in voor beschikbaarheid
1 * 50 mL
Product Details & Documents
10X blocking buffer (pH 7.5) for serum-free blocking of non-specific antibody binding. - Cited in over 30 publications - Suitable for ELISA, in-cell ELISA, ICC/IF, western blot (WB) - 100% casein blocking as preservative
Let op: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specificaties
Specificaties
  • Omschrijving:
    10X Blocking Buffer
Specificaties
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.