About this item
BBP®12 gives you the versatility to print all your basic label types from one machine-saving costs, reducing downtime and simplifying your labelling.
Specifically designed small-core media rolls
Ribbons provide fast installation and reduce change-over time
Centring ribbon spindle virtually eliminates ribbon wrinkle
Compact size won’t clutter your workplace or bench
Over 800 varieties of labels, including external-feed bulk rolls and custom parts, for general identification, wire marking, asset tracking and more.
Ships with a licence for the Brady Workstation suite of your preference: Product and wire identification, safety and facility identification, or lab identification, enabling easy creation of a wide range of labels and signs.
Applications include asset tracking; barcode labelling, circuit board labelling, component and equipment labelling; general identification.
Keyboard type: None
UL; cUL; RoHS; EAC.
THT-; PS-; 3PS-; HX-; 2HX-.
One-year warranty.
It ships with an unwinder, a B-7425 autoclave and liquid nitrogen label, a USB cable, a ribbon R64000, power cord, product CD, quick start guide and a licence for Brady Workstation Software suites.
This BBP®12 label printers comes in a cardboard box.
Specifications
- Catalog No:
- BRDY324280
- BRDY324279
- Aansluitingen:
- Ethernet; Serial; USB
- Ethernet; Serial; USB
- Model:
- BBP®12
- BBP®12
- Power supply:
- EU plug
- UK plug
- Afdrukbreedte:
- 105,41 mm (max.)
- 105,41 mm (max.)
- Breedte:
- 116,84 mm (max, label/tape)
- 116,84 mm (max, label/tape)