Over dit artikel
Create large 38,10 mm labels with edge-to-edge printing and a wide range of text sizes.
Print up to 38,10 mm per second to get work done faster
Backlit full-colour LCD graphics display for easier labelling in low-lit areas
Durably designed to withstand 1,2 m drops
The rechargeable NiMH battery prints up to 1700 labels on a single charge
With 250+ hassle-free drop-in cartridges and 26+ high-performance materials. Enter data with more accuracy as you work between an intuitive QWERTY keypad, a separate numeric keypad or a PC.
Ships with a licence for the brady workstation suite of your preference: product & wire identification, safety & facility identification, or lab identification, enabling easy creation of a wide range of labels and signs.
Applications include asset tracking; circuit board labelling; component and equipment labelling; data and telecommunications labelling; electrical labelling; facility identification; general identification; inventory and inspection labelling; laboratory labelling; lean and 5s labelling; safety identification; warehouse marking; wire and cable labelling; barcode labelling; healthcare and hospital labelling.
Keyboard type: QWERTY
UKCA; FCC.
M4-; M4C-; M5-; M5C-.
Two-year warranty.
This M510 Printer ships in a cardboard box with a rechargeable NiMH battery pack, an AC adapter, B-595 vinyl label cartridge, B-481 polyester slide labels cartridge, a quick start guide, a USB cable, a licence for Brady Workstation Software suites and a USB flash drive with driver and user manual.
Specificaties
- Catalog No:
- BRDY324276
- BRDY324275
- Aansluitingen:
- USB
- USB
- Model:
- M510
- M510
- Power supply:
- EU plug
- UK plug
- Afdrukbreedte:
- 36,58 mm (max.)
- 36,58 mm (max.)
- Breedte:
- 38,10 mm (max, label/tape)
- 38,10 mm (max, label/tape)
Specificaties
Gerelateerde informatie
M510 Label Printer Frequently Asked Questions.pdf
Aanvullende productdetails
Manual M510 Label Printer.pdf
Handmatig
TDS M510, M511 Label Printer.pdf