Order Entry
Belgium
ContactUsLinkComponent
VWR® Wire Rack Bottle Carrier, 6-Place, 80 mm, White
VWR® Wire Rack Bottle Carrier, 6-Place, 80 mm, White
  525-1621
New Product
undefined
VWR® Wire Rack Bottle Carrier, 6-Place, 80 mm, White
  525-1621
New Product

 

  • Capaciteit:
    6 bottles (up to Ø 80 mm)
  • Kleur:
    wit
  • Omschrijving:
    HDPE coated wire rack bottle carrier, autoclavable
  • b×d×h:
    252×174×104 mm

 

 

The VWR® wire rack bottle carrier features a durable, chemical-resistant HDPE-coated wire frame that holds 6 bottles up to 80 mm in diameter. Its swing-down handle allows easy loading and convenient transport. This rack simplifies the process of loading, unloading, and transporting bottles, making it an ideal choice for laboratory and industrial use.

  • HDPE coated wire for durability and chemical resistance

  • Handle design swings down to load and unload without obstruction and swings up for carrying convenience
  • Holds bottles up to 80 mm in diameter

Space saving design.