Orders LinkContactUsLinkComponent
Pipette Tips System, Multirack Advanced
Pipette Tips System, Multirack Advanced
New Product
Leverancier:   RATIOLAB
Pipette Tips System, Multirack Advanced
Leverancier:  RATIOLAB
New Product
Certificaten
Pipette Tips System, Multirack Advanced
2 Opties Hieronder Beschikbaar
Product Details & Documents
The ratiolab® Multiracks advanced provide more time-saving, perfect hygienic conditions and simple operation.
  • Completely autoclavable at 121 °C, 20 minutes
    Durable and steady
    Pipette tips are placed on a robust matrix, in an 8×12 format for all tips
Ahe tips can be picked up by 1, 8 or 12-channel pipettors as well as by pipetting robots contamination-free and without any hand contact. The hinged cover can be opened up to 180°. After work, the Multiracks advanced can be closed firmly through a click closure to protect the tips from dust.
Product Family Options
Productinformatie
  • Tip stijl
    Volume
    Lengte
    Kleurcode
    Voor gebruik met
    Verpakt
    Beschikbaarheid
    Price
  • Pipette tip 'E'
    1 - 200 μl
    49,50 mm
    geel
    ratiopetta®, slamed®, all commonly used pipettes
    Multirack advanced 10×96
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 46,90 €
    46,90 €
    1 * 1 ST
       46,90 €
    Specificaties:
    New Product
    New Product
  • Pipette tip 'E'
    100 - 1200 μl
    70,94 mm
    blauw
    ratiopetta®, slamed®, all commonly used pipettes
    Multirack advanced 10×96
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 49,50 €
    49,50 €
    1 * 1 ST
       49,50 €
    Specificaties:
    New Product