KR2i Starter Fitting Kit
KR2i starter kit
Catalogus # SPECACR2-SKT
New Product
Leverancier: Spectrum Laboratories
Ga naar:
Product Details & Documents
Kit with Luer, mini TC accessories and tools.
Kit consists of fittings: 3 Luer pressure Sensor, non-irradiated (PS), 5 flow control valve (PS), 20 orange cable tie 152,4L×9,52 W mm one way stopcock (nylon), 4 sanitary clamp 12,7×19,05 mm(nylon), 10 sanitary gakset 19,05 mm (silicone), 4 ProConnex®, 19,05 mm TC×6,35 mm HB with FLL (PS), 5 stopcock (PS), 6 ProConnex®, 12,7 mm TC to FLL (PS), 6 Sanitary gasket 12,7 mm (silicone).
Consists of tools: 1 cable tie gun, 1 scissors, 2 cable box and 1 screwdriver (Philips).
Specificaties
- Omschrijving:KR2i starter kit