Tork S5 Clarity Hand Washing Mini Foam Soap
Catalogus # SCAA525202
Leverancier: Essity
New Product
Specificaties
- Verpakt:8 cans per box, 525 ml each
- Omschrijving:Premium foam soap, refill
Specificaties
Over dit artikel
Tork clarity hand washing mini foam soap S5, factory-sealed bottle. 99% of ingredients are from natural origin (according to specification and ISO 16128, including water).
Factory-sealed bottle with a new pump for every refill helps reduce risk of contamination
- Washing ingredients are from renewable sources (vegetable origins) and selected for their lower impact on aquatic life - based upon the EU Ecolabel, CDV (Critical Dilution Volume) related to toxicity to aquatic organisms is below specified limits and is two times lower than the Tork mildly scented foam soap formula
-
The formula is readily biodegradable (based on third part lab test according to OECD301B)
Perfume free fragrance, premium quality
Colour: Transparent
Compatible with S5 dispensers
Sustainability aspirations can be achieved without compromising hand hygiene with the new Tork clarity foam soap. This foam soap is formulated using a limited number of ingredients, 99% from natural origins. It is suitable for Tork mini soap and sanitiser dispensers, certified easy to use and provides good hand hygiene to all users.
Certificeringen:
EU Ecolabel, SE/030/002.