Cryo Gloves and Protective Wear, Thermo Fisher Scientific
Catalog # HERA4426
Supplier: Thermo Scientific
Specifications
- Omschrijving:Cryohandschoenen, large
Specifications
About this item
Keep your hands protected from ultra-low and cryo temperatures with these highly insulated cryo gloves.
Comfortable, lightweight, and breathable protection
- Available in a variety of sizes
Certifications:
cULus and CE.
Packaging:
One pair of cryo gloves provided.