Milli-Q SQPAK™ RO Cartridge
Catalog # MILLSQPAKPR0M1
Supplier: Merck
New Product
Specifications
- Omschrijving:SQPAK™ RO Cartridge, removes organics, ions, particulates up to 99% from tap water feeding Milli-Q® SQ 2Series RO production stations
Specifications
About this item
The SQPAK™ RO cartridge produces pure RO water at up to 42 L/h. This cartridge′s RO membrane efficiently removes up to 99% of water contaminants such as bacteria, ions, particulates and organics.
- Efficiently removes up to 99% of ions, bacteria, particulates and organics from feed water
- Easy to replace thanks to the Twist and Lock mechanism
- Need for cartridge replacement is indicated by an orange, blinking pictogram light on the system
Pure label and green colour of the SQPAK™ RO cartridge is repeated on the production station interface, to inform about its water purification purpose and guide replacement.