Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Human Tissue Factor ELISA Kit (CD142)

Human Tissue Factor ELISA Kit (CD142)

Human Tissue Factor ELISA kit (CD142), 96 tests
Catalogus # ABCAAB108903-96
Leverancier:   Abcam
Human Tissue Factor ELISA Kit (CD142)
Catalogus # ABCAAB108903-96
Leverancier:  Abcam
CAS-nummer:  
Human Tissue Factor ELISA Kit (CD142)
Human Tissue Factor ELISA Kit (CD142)

Human Tissue Factor ELISA kit (CD142), 96 tests

CAS-nummer:  
Price
805,00 €
Log in voor beschikbaarheid
1 * 96 Tests
Product Details & Documents

Human Tissue Factor ELISA kit (CD142) is a Sandwich (quantitative) ELISA kit for the measurement of Human Tissue Factor (CD142) in Human in plasma, cell culture extracts, Tissue, Serum samples.

Specificaties

Specificaties

  • Omschrijving:
    Human Tissue Factor ELISA kit (CD142)
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Formaat:
    Pre-coated
  • Assaytype:
    Sandwich
  • Time to Results:
    5 h 0 min
  • Monstertype:
    Plasma, cell culture extracts, tissue, serum
  • Cat. No.:
    ABCAAB108903-96
  • Gevoeligheid:
    20 pg/ml
  • Opslagtemperatuur:
    4 °C
  • Target protein:
    Tissue Factor
  • Detectiemethode:
    Colorimetric
  • Detectiebereik:
    12,5 - 100 pg/ml
  • Duur van de test:
    Multiple steps
  • Regulatory status:
    RUO

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.