Digital thermo-hygrometer, TFH 620
Catalog # 620-1394
Supplier: EBRO
Specifications
- Omschrijving:TFH 620 with air probe TPH 100
- Juistheid:±0,3 °C; ±2,0 % HR
- VPE:1
Specifications
About this item
This robust and impact resistant thermo-hygrometer provides high accuracy measurements for monitoring storage checks.
- Automatic deactivation
- Capacitive humidity sensor
- Two measurement channels
Thermistor sensor.
Delivery: Supplied with an internal sensor, a six-point factory calibration certificate, a lithium three-volt battery for up to five years of life with a charge indicator.