Laboratory tables
With 4 swivel castors, 2 of them with brake in galvanised-chromed housing, Ø 125 mm, 450×450×510 mm
Catalogus # HUPF0114932
Leverancier: HUPFER METALLWERKE
Ga naar:
Product Details & Documents
Working table is mobile and made of KHfa/45 stainless steel.
Specificaties
- Omschrijving:With 4 swivel castors, 2 of them with brake in galvanised-chromed housing, Ø 125 mm
- b×d×h:450×450×510 mm