Orders LinkContactUsLinkComponent
Microtubes, with Thread
Marketsource
Leverancier:   SARSTEDT
Microtubes, with Thread
Leverancier:  SARSTEDT
Marketsource
Certificaten
25 Opties Hieronder Beschikbaar
Product Details & Documents
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.
Product Family Options
Tubes, 0,5 ml, with skirted base, neutral screw cap
Productinformatie
  • Capaciteit
    Verpakt
    Aanbevolen werkvolume
    Beschikbaarheid
    Price
  • SARS72.730.006
    Micro tubes with neutral screw cap assembled, with writing space, sterile, skirted base
    Micro tubes with neutral screw cap assembled, with writing space, sterile, skirted base
    0,5 ml
    1000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 166,66 €
    166,66 €
    1 * 1.000 ST
       166,66 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.730.005
    Micro tubes with neutral screw cap assembled, sterile, skirted base
    Micro tubes with neutral screw cap assembled, sterile, skirted base
    0,5 ml
    1000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 100,18 €
    100,18 €
    1 * 1.000 ST
       100,18 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.730
    Micro tubes with neutral screw cap enclosed, skirted base
    Micro tubes with neutral screw cap enclosed, skirted base
    0,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 551,45 €
    551,45 €
    1 * 5.000 ST
       551,45 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.730.003
    Micro tubes without screw cap, skirted base
    Micro tubes without screw cap, skirted base
    0,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 290,80 €
    290,80 €
    1 * 5.000 ST
       290,80 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.730.004
    Micro tubes, amber, without screw cap, skirted base
    Micro tubes, amber, without screw cap, skirted base
    0,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 226,14 €
    226,14 €
    1 * 5.000 ST
       226,14 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.730.007
    Micro tubes with neutral screw cap enclosed, with writing space, skirted base
    Micro tubes with neutral screw cap enclosed, with writing space, skirted base
    0,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 542,62 €
    542,62 €
    1 * 5.000 ST
       542,62 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.730.009
    Micro tubes with writing space, without screw cap, skirted base
    Micro tubes with writing space, without screw cap, skirted base
    0,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 248,07 €
    248,07 €
    1 * 5.000 ST
       248,07 €
    Specificaties:
    Marketsource
    Marketsource
Tubes, 0,5 ml, without skirted base, neutral screw cap
Productinformatie
  • Capaciteit
    Verpakt
    Aanbevolen werkvolume
    Beschikbaarheid
    Price
  • SARS72.733.001
    Micro tubes with neutral screw cap enclosed, without skirted base
    Micro tubes with neutral screw cap enclosed, without skirted base
    0,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 410,26 €
    410,26 €
    1 * 5.000 ST
       410,26 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.733.010
    Micro tubes without screw cap, without skirted base
    Micro tubes without screw cap, without skirted base
    0,5 ml
    5000/case
    0,5 ml
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 374,94 €
    374,94 €
    1 * 5.000 ST
       374,94 €
    Specificaties:
    Marketsource
    Marketsource
Tubes, 1,5 ml, with skirted base, without screw cap
Productinformatie
  • Capaciteit
    Verpakt
    Aanbevolen werkvolume
    Beschikbaarheid
    Price
  • SARS72.703
    Micro tubes without screw cap, skirted base
    Micro tubes without screw cap, skirted base
    1,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 213,25 €
    213,25 €
    1 * 5.000 ST
       213,25 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.703.004
    Micro tubes, amber, without screw cap, skirted base
    Micro tubes, amber, without screw cap, skirted base
    1,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 226,04 €
    226,04 €
    1 * 5.000 ST
       226,04 €
    Specificaties:
    Marketsource
    Marketsource
Tubes, 1,5 ml, without skirted base, with neutral screw cap
Productinformatie
  • Capaciteit
    Verpakt
    Aanbevolen werkvolume
    Beschikbaarheid
    Price
  • SARS72.692
    Micro tubes with neutral screw cap enclosed, without skirted base
    Micro tubes with neutral screw cap enclosed, without skirted base
    1,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 291,10 €
    291,10 €
    1 * 5.000 ST
       291,10 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.692.005
    Micro tubes with neutral screw cap assembled, sterile, without skirted base
    Micro tubes with neutral screw cap assembled, sterile, without skirted base
    1,5 ml
    1000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 129,41 €
    129,41 €
    1 * 1.000 ST
       129,41 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.607
    Micro tubes without screw cap, without skirted base
    Micro tubes without screw cap, without skirted base
    1,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 211,73 €
    211,73 €
    1 * 5.000 ST
       211,73 €
    Specificaties:
    Marketsource
    Marketsource
Tubes, without knurls, with or without neutral screw cap
Productinformatie
  • Capaciteit
    Verpakt
    Aanbevolen werkvolume
    Beschikbaarheid
    Price
  • SARS72.664
    Micro tubes with skirted base, without screw cap, without knurls
    Micro tubes with skirted base, without screw cap, without knurls
    2,0 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 121,19 €
    121,19 €
    1 * 5.000 ST
       121,19 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.687
    Micro tubes without skirted base, without screw cap, without knurls
    Micro tubes without skirted base, without screw cap, without knurls
    1,5 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 121,19 €
    121,19 €
    1 * 5.000 ST
       121,19 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.785.005
    Micro tubes with skirted base, neutral screw cap assembled, sterile, without knurls
    Micro tubes with skirted base, neutral screw cap assembled, sterile, without knurls
    0,5 ml
    1000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 143,01 €
    143,01 €
    1 * 1.000 ST
       143,01 €
    Specificaties:
    Marketsource
    Marketsource
Tubes, 2 ml, without skirted base, neutral screw cap
Productinformatie
  • Capaciteit
    Verpakt
    Aanbevolen werkvolume
    Beschikbaarheid
    Price
  • SARS72.693
    Micro tubes with neutral screw cap enclosed, without skirted base
    Micro tubes with neutral screw cap enclosed, without skirted base
    2,0 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 302,27 €
    302,27 €
    1 * 5.000 ST
       302,27 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.693.005
    Micro tubes with neutral screw cap assembled, sterile, without skirted base
    Micro tubes with neutral screw cap assembled, sterile, without skirted base
    2,0 ml
    1000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 129,41 €
    129,41 €
    1 * 1.000 ST
       129,41 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.608
    Micro tubes without screw cap, without skirted base
    Micro tubes without screw cap, without skirted base
    2,0 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 211,73 €
    211,73 €
    1 * 5.000 ST
       211,73 €
    Specificaties:
    Marketsource
    Marketsource
Tubes, 2 ml, with skirted base, neutral screw cap
Productinformatie
  • Capaciteit
    Verpakt
    Aanbevolen werkvolume
    Beschikbaarheid
    Price
  • SARS72.694.005
    Micro tubes with neutral screw cap assembled, sterile, with skirted base
    Micro tubes with neutral screw cap assembled, sterile, with skirted base
    2,0 ml
    1000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 129,41 €
    129,41 €
    1 * 1.000 ST
       129,41 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.609
    Micro tubes without screw cap, with skirted base
    Micro tubes without screw cap, with skirted base
    2,0 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 211,73 €
    211,73 €
    1 * 5.000 ST
       211,73 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.694.006
    Micro tubes with writing space and graduation, neutral screw cap assembled, sterile, with skirted base
    Micro tubes with writing space and graduation, neutral screw cap assembled, sterile, with skirted base
    2,0 ml
    1000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 153,98 €
    153,98 €
    1 * 1.000 ST
       153,98 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.609.003
    Micro tubes, amber, without screw cap, with skirted base
    Micro tubes, amber, without screw cap, with skirted base
    2,0 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 224,21 €
    224,21 €
    1 * 5.000 ST
       224,21 €
    Specificaties:
    Marketsource
    Marketsource
  • SARS72.609.001
    Micro tubes with writing space and graduation, without screw cap, with skirted base
    Micro tubes with writing space and graduation, without screw cap, with skirted base
    2,0 ml
    5000/case
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 5.000 ST
    1 * 5.000 ST - 332,31 €
    332,31 €
    1 * 5.000 ST
       332,31 €
    Specificaties:
    Marketsource
    Marketsource
Screw caps for micro tubes
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes
    zwart
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 54,63 €
    54,63 €
    1 * 1.000 ST
       54,63 €
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes
    violet
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 54,63 €
    54,63 €
    1 * 1.000 ST
       54,63 €
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes
    wit
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 10.000 ST
    1 * 10.000 ST - 546,27 €
    546,27 €
    1 * 10.000 ST
       546,27 €
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes, 10 colours
    gesordeerd
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 10.000 ST
    1 * 10.000 ST - 380,22 €
    380,22 €
    1 * 10.000 ST
       380,22 €
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes
    natuur
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 54,63 €
    54,63 €
    1 * 1.000 ST
       54,63 €
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes
    groen
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 10.000 ST
    1 * 10.000 ST - 54,63 €
    54,63 €
    1 * 10.000 ST
       54,63 €
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes
    oranje
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 54,63 €
    54,63 €
    1 * 1.000 ST
       54,63 €
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes
    blauw
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 54,63 €
    54,63 €
    1 * 1.000 ST
       54,63 €
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes
    rood
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 54,63 €
    54,63 €
    1 * 1.000 ST
       54,63 €
Productinformatie
  • Catalogus #
    Omschrijving
    Kleur
    Beschikbaarheid
    Price
  • Screw caps for micro tubes
    roos
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1.000 ST
    1 * 1.000 ST - 54,63 €
    54,63 €
    1 * 1.000 ST
       54,63 €