Orders LinkContactUsLinkComponent
Transfer pipettes
Marketsource
 :   SARSTEDT
Transfer pipettes
 :  SARSTEDT
Marketsource
Transfer pipettes
LDPE, disposable.
  • No risk of injury or infection from broken glass
  • Disposable: No carry over or cross contamination
  • Can be heat-sealed for sample storage and transport
  • Environmentally neutral, ensuring non-toxic disposal
Product Family Options
Non sterile
  • Omschrijving
    Capaciteit
    Druppelvolume
    Lengte
    Schaalverdeling
    Steriel
    Verpakt
  • Transferpipet, LDPE, type I, voor eenmalig gebruik
    1,0 ml
    30 - 45 µl
    87 mm
    - ml
    -
    Bulk
    1 * 3.000 ST
    1 * 3.000 ST - 174,09 €
    174,09 €
    1 * 3.000 ST
       174,09 €
    Marketsource
    Marketsource
  • 1,0 ml
    17 - 22 µl
    104
    - ml
    -
    Bulk
    1 * 3.000 ST
    1 * 3.000 ST - 174,09 €
    174,09 €
    1 * 3.000 ST
       174,09 €
    Marketsource
    Marketsource
  • 1,0 ml
    45 - 55 µl
    115 mm
    0,10 ml
    -
    Bulk
    1 * 2.000 ST
    1 * 2.000 ST - 116,08 €
    116,08 €
    1 * 2.000 ST
       116,08 €
    Marketsource
    Marketsource
  • 2,0 ml
    35 - 45 µl
    154 mm
    - ml
    -
    Bulk
    1 * 3.000 ST
    1 * 3.000 ST - 174,09 €
    174,09 €
    1 * 3.000 ST
       174,09 €
    Marketsource
    Marketsource
  • Transferpipet, LDPE, type D, voor eenmalig gebruik
    3,5 ml
    30 - 45 µl
    155 mm
    - ml
    -
    Bulk
    1 * 2.000 ST
    1 * 2.000 ST - 99,35 €
    99,35 €
    1 * 2.000 ST
       99,35 €
    Marketsource
    Marketsource
  • 3,5 ml
    30 - 45 µl
    156 mm
    0,25 ml
    -
    Bulk
    1 * 2.000 ST
    1 * 2.000 ST - 91,59 €
    91,59 €
    1 * 2.000 ST
       91,59 €
    Marketsource
    Marketsource
  • 3,5 ml
    35 - 55 µl
    155 mm
    0,50 ml
    -
    Bulk
    1 * 2.000 ST
    1 * 2.000 ST - 92,20 €
    92,20 €
    1 * 2.000 ST
       92,20 €
    Marketsource
    Marketsource
  • 6,0 ml
    20 - 30 µl
    146 mm
    - ml
    -
    Bulk
    1 * 1.600 ST
    1 * 1.600 ST - 95,29 €
    95,29 €
    1 * 1.600 ST
       95,29 €
    Marketsource
    Marketsource
  • 6,0 ml
    40 - 55 µl
    152 mm
    - ml
    -
    Bulk
    1 * 1.600 ST
    1 * 1.600 ST - 72,47 €
    72,47 €
    1 * 1.600 ST
       72,47 €
    Marketsource
    Marketsource
Sterile
  • Omschrijving
    Capaciteit
    Druppelvolume
    Lengte
    Schaalverdeling
    Steriel
    Verpakt
  • 3,5 ml
    35 - 55 µl
    155 mm
    0,50 ml
    +
    10 per zak
    1 * 1.200 ST
    1 * 1.200 ST - 94,03 €
    94,03 €
    1 * 1.200 ST
       94,03 €
    Marketsource
    Marketsource
  • 3,5 ml
    35 - 55 µl
    155 mm
    0,50 ml
    +
    Afzonderlijk verpakt
    1 * 840 ST
    1 * 840 ST - 97,44 €
    97,44 €
    1 * 840 ST
       97,44 €
    Marketsource
    Marketsource
  • 3,5 ml
    35 - 45 µl
    156 mm
    0,25 ml
    +
    Afzonderlijk verpakt
    1 * 840 ST
    1 * 840 ST - 97,44 €
    97,44 €
    1 * 840 ST
       97,44 €
    Marketsource
    Marketsource
  • 6,0 ml
    20 - 30 µl
    146 mm
    - ml
    +
    10 per zak
    1 * 1.000 ST
    1 * 1.000 ST - 119,77 €
    119,77 €
    1 * 1.000 ST
       119,77 €
    Marketsource
    Marketsource
  • 6,0 ml
    20 - 30 µl
    146 mm
    - ml
    +
    Afzonderlijk verpakt
    1 * 840 ST
    1 * 840 ST - 105,76 €
    105,76 €
    1 * 840 ST
       105,76 €
    Marketsource
    Marketsource