Molecular models: RNA, Molymod®
Marketsource
Leverancier: SPIRING ENTERPRISES
Ga naar:
Over dit artikel
The Molymod system is the original dual-scale system of high quality molecular models.
Levering: Protein synthesis kit with: 24 bases, 12 ribose/phophate, 4 TRNA, 4 amino acid parts.
2 Opties Hieronder Beschikbaar
Product Details & Documents
The Molymod system is the original dual-scale system of high quality molecular models.
Levering: Protein synthesis kit with: 24 bases, 12 ribose/phophate, 4 TRNA, 4 amino acid parts.
Product Family Options
Productinformatie
- BeschikbaarheidPrice
- SPIRRNA-24Molecular model of RNASpecificaties:MarketsourceMarketsource
- SPIRMRNA-24-PDMolecular model of mRNASpecificaties:MarketsourceMarketsource