Tanks
Leverancier: ROETZMEIER SICHERH BEHAELTER
Ga naar:
Over dit artikel
2 Opties Hieronder Beschikbaar
Product Family Options
Productinformatie
- CapaciteitTypeBeschikbaarheidPrice
- 119-0154Washing tank for safe cleaning of particels and tools with inflammable liquidsWashing tank for safe cleaning of particels and tools with inflammable liquids4 lWastankLog in voor beschikbaarheid1 * 1 ST57,00 €
- 119-0143Soaking tank for controlled moistening, soaking or bathing of particels, cloths, flannels, paper, etc., e.g. for cleaning with solventsSoaking tank for controlled moistening, soaking or bathing of particels, cloths, flannels, paper, etc., e.g. for cleaning with solvents4 lInweektankLog in voor beschikbaarheid1 * 1 ST209,00 €