Solid phase extraction cartridges, Bond Elut™ SCX
Over dit artikel
Bond Elut™ SCX is a strong cation exchanger with a very low pKa. Although the pKa is similar to Bond Elut™ PRS, the presence of the benzene ring in the functional group increases the potential for non-polar interactions. This non-polar characteristic becomes particularly important when conducting ion-exchange from aqueous systems, where selectivity towards compounds exhibiting cationic and non-polar character is seen.
- Useful for compounds with both cationic and non-polar characteristics
- Superior cleanup from a single sorbent
- Very low pKa ligand elicits strong analyte interaction
24 Opties Hieronder Beschikbaar
Product Details & Documents
Bond Elut™ SCX is a strong cation exchanger with a very low pKa. Although the pKa is similar to Bond Elut™ PRS, the presence of the benzene ring in the functional group increases the potential for non-polar interactions. This non-polar characteristic becomes particularly important when conducting ion-exchange from aqueous systems, where selectivity towards compounds exhibiting cationic and non-polar character is seen.
- Useful for compounds with both cationic and non-polar characteristics
- Superior cleanup from a single sorbent
- Very low pKa ligand elicits strong analyte interaction
Specifications for Product Family
Specificaties
- Catalog No:
- VARI12113013
- 551-0347
- VARI14113039
- 549-0791
- 551-0309
- 549-0797
- 551-0323
- 549-0821
- 549-0833
- 549-0834
- 549-0871
- 549-0852
- 549-0842
- 549-0847
- 549-0857
- 549-0862
- 549-0867
- 551-0376
- 551-0383
- 551-0431
- VARI75507050
- VARI7550701C
- VARI7550702C
- VARI7550704C
- Koolstofgehalte:
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- 10,9%
- Endcapping:
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Neen
- Oppervlakte:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
Specificaties
Product Family Options
LRC cartridges
Productinformatie
- FasePoriëngroottePartikelgrootteCapaciteitBedgewichtBeschikbaarheidPrice
- VARI12113013LRC-patronenSpecificaties:
More
Specifications
Cat. No.VARI12113013Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 551-0347LRC-patronenSpecificaties:
More
Specifications
Cat. No.551-0347Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - VARI14113039LRC-patronenSpecificaties:
More
Specifications
Cat. No.VARI14113039Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g
Straight barrel cartridges
Productinformatie
- FasePoriëngroottePartikelgrootteCapaciteitBedgewichtBeschikbaarheidPrice
- 549-0791Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0791Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 551-0309Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.551-0309Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0797Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0797Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 551-0323Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.551-0323Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0821Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0821Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0833Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0833Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0834Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0834Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0871Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0871Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0852Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0852Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0842Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0842Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0847Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0847Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0857Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0857Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0862Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0862Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 549-0867Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.549-0867Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g
Bond Elut™ Jr
Productinformatie
- FasePoriëngroottePartikelgrootteCapaciteitBedgewichtBeschikbaarheidPrice
- 551-0376Bond Elut™ JrSpecificaties:
More
Specifications
Cat. No.551-0376Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - 551-0383Bond Elut™ JrSpecificaties:
More
Specifications
Cat. No.551-0383Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g
VersaPlate™ formats
Productinformatie
- FasePoriëngroottePartikelgrootteCapaciteitBedgewichtBeschikbaarheidPrice
- 551-0431Voorgemonteerde plaat met 96 putjesSpecificaties:
More
Specifications
Cat. No.551-0431Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - VARI75507050VersaPlate™-buisSpecificaties:
More
Specifications
Cat. No.VARI75507050Koolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - VARI7550701CVersaPlate™-buisSpecificaties:
More
Specifications
Cat. No.VARI7550701CKoolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - VARI7550702CVersaPlate™ well plate tubesSpecificaties:
More
Specifications
Cat. No.VARI7550702CKoolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g - VARI7550704CVersaPlate™ well plate tubesSpecificaties:
More
Specifications
Cat. No.VARI7550704CKoolstofgehalte10,9%EndcappingNeenOppervlakte500 m²/g
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.