Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ DEA
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ DEA
Supplier:  VARIAN
Certificates
11 Options Available Below
Product Details & Documents
Bond Elut™ DEA bears some resemblance to Bond Elut™ NH₂ in its properties but with a slightly lower capacity as an anion exchange sorbent. DEA has a moderately non-polar character due to the alkyl side chains on the amino functionality. These groups still afford a medium level of polarity, higher than C₈ but less polar than C₂ or CN-E.
  • More polar than C₈ but less polar than C₂ or CN
  • Lower ionic capacity than NH₂
  • Alkyl side chains confer moderately non-polar chracteristics
Bond Elut™ DEA is an excellent phase for the extraction of bisphosphonates from human serum and urine. Bisphosphonates are extremely hydrophilic and structurally similar to many endogenous phosphorylated compounds, making their selective extraction from serum or urine very challenging. The unique chemistry of Bond Elut™ DEA offers excellent selectivity of this class of compounds.
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12113016
  • VARI12113042
  • VARI14113042
  • VARI12102043
  • VARI12102016
  • VARI14102078
  • VARI14102016
  • VARI14102043
  • VARI102019
  • VARI12102078
  • VARI12166046B
  • Koolstofgehalte:
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • 8,5%
  • Endcapping:
  • Neen
  • Neen
  • Neen
  • Neen
  • Neen
  • Neen
  • Neen
  • Neen
  • Neen
  • Neen
  • Neen
  • Oppervlakte:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
  • Omschrijving
    Fase
    Poriëngrootte
    Partikelgrootte
    Capaciteit
    Bedgewicht
    Availability
    Price
  • LRC-patronen
    DEA
    60 Å
    40 µm
    10 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 173,00 €
    173,00 €
    1 * 50 ST
       173,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113016
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
  • LRC-patronen
    DEA
    60 Å
    40 µm
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 243,00 €
    243,00 €
    1 * 50 ST
       243,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12113042
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
  • LRC-patronen
    DEA
    60 Å
    120 µm
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 243,00 €
    243,00 €
    1 * 50 ST
       243,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14113042
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
Straight barrel cartridges
Product Information
  • Omschrijving
    Fase
    Poriëngrootte
    Partikelgrootte
    Capaciteit
    Bedgewicht
    Availability
    Price
  • Straight barrel cartridge
    DEA
    60 Å
    120 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 261,00 €
    261,00 €
    1 * 50 ST
       261,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102043
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
  • Straight barrel cartridge
    DEA
    60 Å
    40 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 301,00 €
    301,00 €
    1 * 100 ST
       301,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102016
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
  • Straight barrel cartridge
    DEA
    60 Å
    120 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102078
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
  • Straight barrel cartridge
    DEA
    60 Å
    40 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 301,00 €
    301,00 €
    1 * 100 ST
       301,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102016
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
  • Straight barrel cartridge
    DEA
    60 Å
    40 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 261,00 €
    261,00 €
    1 * 50 ST
       261,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI14102043
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
  • Straight barrel cartridge
    DEA
    60 Å
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
    Specifications:
    More
    Specifications
    Cat. No.
    VARI102019
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
  • Straight barrel cartridge
    DEA
    60 Å
    40 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 293,00 €
    293,00 €
    1 * 100 ST
       293,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12102078
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
Bond Elut™ Jr
Product Information
  • Omschrijving
    Fase
    Poriëngrootte
    Partikelgrootte
    Capaciteit
    Bedgewicht
    Availability
    Price
  • Bond Elut™ Jr
    DEA
    60 Å
    40 µm
    1000 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 402,00 €
    402,00 €
    1 * 100 ST
       402,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12166046B
    Koolstofgehalte
    8,5%
    Endcapping
    Neen
    Oppervlakte
    500 m²/g
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.