Solid phase extraction cartridges, Bond Elut™ C2
Over dit artikel
Bond Elut™ C2 is a fairly non-polar sorbent because of the short chain length of the functional group. C2 is often used during the process of method development if analytes are retained too strongly on a C8 or C18 phase. The polarity of C2 is slightly lower than a cyano phase for polar interactions.
- C2 is a low carbon load sorbent
- Can be used alongside CN and C8 phases
- Popular for drug extraction from plasma and for flat baselines
10 Opties Hieronder Beschikbaar
Product Details & Documents
Bond Elut™ C2 is a fairly non-polar sorbent because of the short chain length of the functional group. C2 is often used during the process of method development if analytes are retained too strongly on a C8 or C18 phase. The polarity of C2 is slightly lower than a cyano phase for polar interactions.
- C2 is a low carbon load sorbent
- Can be used alongside CN and C8 phases
- Popular for drug extraction from plasma and for flat baselines
Specifications for Product Family
Specificaties
- Catalog No:
- VARI12113003
- VARI12113029
- VARI12102060
- VARI12105029
- 551-0302
- VARI12102117
- 551-0314
- 551-0317
- VARI12102115
- VARI12256003
- Koolstofgehalte:
- 5,6%
- 5,6%
- 5,6%
- 5,6%
- 5,6%
- 5,6%
- 5,6%
- 5,6%
- 5,6%
- 5,6%
- Endcapping:
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Oppervlakte:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
Specificaties
Product Family Options
LRC cartridges
Productinformatie
- FasePoriëngroottePartikelgrootteCapaciteitBedgewichtBeschikbaarheidPrice
- VARI12113003LRC-patronenSpecificaties:
More
Specifications
Cat. No.VARI12113003Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g - VARI12113029LRC-patronenSpecificaties:
More
Specifications
Cat. No.VARI12113029Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g
Straight barrel cartridges
Productinformatie
- FasePoriëngroottePartikelgrootteCapaciteitBedgewichtBeschikbaarheidPrice
- VARI12102060Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.VARI12102060Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g - VARI12105029Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.VARI12105029Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g - 551-0302Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.551-0302Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g - VARI12102117Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.VARI12102117Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g - 551-0314Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.551-0314Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g - 551-0317Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.551-0317Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g - VARI12102115Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.VARI12102115Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g - VARI12256003Straight barrel cartridgeSpecificaties:
More
Specifications
Cat. No.VARI12256003Koolstofgehalte5,6%EndcappingJaOppervlakte500 m²/g