Orders LinkContactUsLinkComponent
PIG® leak diverter quick-deploy kit
PIG® leak diverter quick-deploy kit
Leverancier:   NEW PIG
PIG® leak diverter quick-deploy kit
Leverancier:  NEW PIG
Certificaten
PIG® leak diverter quick-deploy kit
PIG® leak diverter quick-deploy kit
2 Opties Hieronder Beschikbaar
Product Details & Documents
Quick deploying leak diverter springs open fast to catch roof leaks. Simply pop it open.
  • Superfast to control leaks and protect people, inventory, and equipment
  • Hangs easily in problem spots to catch nuisance leaks and channel them to a floor drain or collection container
  • Nonpermeable tarp is vinyl coated for strength and durability; resists punctures, UV and mildew for long life
  • Pinched corners help funnel liquid toward center for better drainage
  • Connect flexible PVC drain hose (included) to channel liquid to floor drain or collection container
3/4" drain fitting accepts longer hoses if necessary. Flow rate of 12,6 litres per minute when used with a 1/2" ID hose.
Levering: Kit includes leak diverter, quick connect drain fitting (Ø 1905 mm) and hose of length 3 m.
Product Family Options
Productinformatie
  • l×b
    Beschikbaarheid
    Price
  • NEWPTLS903
    Leak diverter quick-deploy kit
    Leak diverter quick-deploy kit
    1230×1230 mm
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 344,00 €
    344,00 €
    1 * 1 ST
       344,00 €
  • NEWPTLS461-YW
    Leak diverter quick-deploy kit
    Leak diverter quick-deploy kit
    760×760 mm
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 255,00 €
    255,00 €
    1 * 1 ST
       255,00 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.