2-Methoxycyclohexanone ≥95.0% (by GC)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:7429-44-9
:
Formule:C₇H₁₂O₂
MDL-nummer:MFCD00009610
:128.17 g/mol
Kokend Pt:88°C
Dichtheid:1,02 g/cm³ (20 °C)
Flits Pt:69°C
EINECS:231-071-8
Specificatie testresultaten
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specificatie testresultaten
Frequently Bought Together