Stoppers, rubber, conical
17,0 mm
Catalog # HECH40955010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Rubber stopper for centrifuge tubes 10 to 15 ml.
Specifications
- Ø onder:17,0 mm