ContactUsLinkComponent
Accessories for safety dispensing can, stainless steel
Accessories for safety dispensing can, stainless steel
Catalogus # DUEH1.80.000
Leverancier:  DUPERTHAL
undefined
Accessories for safety dispensing can, stainless steel
Catalogus # DUEH1.80.000
Leverancier:  DUPERTHAL
Specificaties
  • Omschrijving:
    Tap, self-closing, 3/4" threaded, sealing material teflon
  • VPE:
    1
Specificaties
Over dit artikel