Blood collection tubes with KF+Na2EDTA
Catalog # KIMA18506
Supplier: KIMA SAS DI CHIARIN RENZO
Specifications
- Ø:12 mm
- Omschrijving:Blood Collection tube
- Lengte:86 mm
- Materiaal:PP
- Kleur dop:oranje
- VPE:10.000
- Additief:KF+Na2EDTA
Specifications
About this item
PP, translucent.
- KF+Na2 EDTA for 2,5ml of blood
- Non sterile
- With writing area