Nissel tubes
Catalog # HECH41145010
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Omschrijving:Tube Nissel (3 ml), with blue graduation, for the volumetric determination of fibrin
- VPE:1
Specifications
About this item
Glass.
- Fused blue graduation
For the volumetric determination of fibrin. With directions for use - "heat fibrin according to Schulz".