Orders LinkContactUsLinkComponent
Holders for test tubes, wood
Holders for test tubes, wood
Catalogus # 241-0416
Leverancier:   HERENZ HEINZ
Holders for test tubes, wood
Catalogus # 241-0416
Leverancier:  HERENZ HEINZ
Holders for test tubes, wood
Holders for test tubes, wood
Price
2,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Test tube holders made of wood.
  • Peg/clip shape
Suitable for tubes up to 20 mm diameter.