Orders LinkContactUsLinkComponent
Staining jars acc.to Hellendahl
Staining jars acc.to Hellendahl
Staining jar, Hellendahl type, 58×53,5×86 mm
Catalog # LPIT290417
Supplier:   LP ITALIANA
Staining jars acc.to Hellendahl
Catalog # LPIT290417
Supplier:  LP ITALIANA
Staining jars acc.to Hellendahl
Staining jars acc.to Hellendahl
Staining jar, Hellendahl type, 58×53,5×86 mm
Price
86,40 €
Log in for availability
1 * 4 ST
Product Details & Documents
Jar made of TPX.
It hold up to 16 microscopic slides.
Specifications
  • Omschrijving:
    Staining jar, Hellendahl type
  • b×d×h:
    58×53,5×86 mm