Orders LinkContactUsLinkComponent
Tripods
Tripods
Tripod, outer Ø 140 mm, 110×280 mm, Branders nr. 1395 en 1396
Catalog # HECH41091010
Tripods
Catalog # HECH41091010
Tripods
Tripods
Tripod, outer Ø 140 mm, 110×280 mm, Branders nr. 1395 en 1396
Price
24,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
These tripods are made of galvanised iron.
Specifications
  • Voor:
    Branders nr. 1395 en 1396
  • Ø inw.×h:
    110×280 mm
  • Omschrijving:
    Tripod, outer Ø 140 mm
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Info VPE
    VPE
    Availability
    Price
  • HECH51071012
    120x120 mm
    1
    Log in for availability
    1 * 1 ST - 3,20 €