Surgical blades for cervical biopsy
Catalogus # SWAN2001
Leverancier: SWANN MORTON
Specificaties
- Voor:Grote S/L-handvatten
- Omschrijving:Baarmoederhalsbiopsiemes, roestvrij staal, verpakt in afzonderlijke plastic laden
- VPE:10
Specificaties
Over dit artikel
This sterile stainless cervical biposy blade is used in conjunction with the long stainless steel Major handle to take a 'cone' biopsy from the tissue in a woman's cervix. The tissue sample can then be analysed to assess the prescence of malignant and non malignant carcinoma cells.
- Fits handles Major S and L