Orders LinkContactUsLinkComponent
Bottles, water sampling
Bottles, water sampling
500 ml, single wrapped
Catalogus # 300-0161
Leverancier:   LP ITALIANA
Bottles, water sampling
Catalogus # 300-0161
Leverancier:  LP ITALIANA
Bottles, water sampling
Bottles, water sampling
500 ml, single wrapped
Price
405,00 €
Log in voor beschikbaarheid
1 * 120 ST
Product Details & Documents
PP, translucent, square, with screw cap.
  • Tamper evident screw caps
  • Sterile
  • Moulded graduations
Bottles for microbiological analysis of chlorinated or non-chlorinated water. Sodium thiosulphate inhibits the effects of chlorine, bromide and ozone present in the water. Ideal for flat storage (fridges, etc).
Caution: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Specificaties
  • Verpakt:
    single wrapped
  • Capaciteit:
    500 ml
  • Seq:
    04