Capillary columns, Elite-Betacydex for autosystem
Catalog # PERCN9316319
Supplier: PerkinElmer
Specifications
- Kolomtype:Capillair
- Filmdikte:0,25 µm
- Ø inw.:0,25 mm
- Lengte:30 m
- Environmentally Preferable:
- Fase:Elite-Betacydex
- VPE:1
Specifications
About this item
Elite-Betacydex columns are ideal for chiral separations.