Orders LinkContactUsLinkComponent
Containers with pressure cap
Containers with pressure cap
Containers with pressure cap
Containers with pressure cap
Container made of PS, clear, with PE snap cap.
  • Good chemical resistance
  • Lightweight
  • Easy to clean
Ideal for collecting, storing and transporting samples.
 : *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Product Family Options
  • Capaciteit
    Ø uitw.
    Hoogte
    Kleur dop
    Uitvoering
    Materiaal
  • 30 ml
    34 mm
    41 mm
    geel
    sterile (ionising radiation)
    1 * 1.000 ST
    1 * 1.000 ST - 359,00 €
    359,00 €
    1 * 1.000 ST
       359,00 €
  • 45 ml
    34 mm
    60 mm
    wit
    non sterile
    PS
    1 * 750 ST
    1 * 750 ST - 166,00 €
    166,00 €
    1 * 750 ST
       166,00 €