Flasks, pear shaped, Claisen-Vigreux, with two necks
Catalog # WITG0.786.025
Supplier: witeg Labortechnik
Specifications
- Capaciteit:25 ml
- VPE:1
- Zijhalzen:14/23 NS
- Middenhals:14/23 NS
Specifications
About this item
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Incorporates a short, Vigreux type, fractionating column. Column indents aid intimate contact between vapour and liquid.