Octaethylene glycol monododecylether (C12E8) 10% in aqueous solution, Calbiochem®
Catalogus # 205532-1
Leverancier: Merck Millipore (Calbiochem)
CAS-nummer:
Specificaties
- VPE:1 SET
- Seq:0125
Specificaties
Over dit artikel
:
:538.754 g/mol
CAS-nummer:3055-98-9
Formule:C₂₈H₅₈O₉
MDL-nummer:MFCD00043234